missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Pulmonary surfactant-associated protein A1 Protein
A DNA sequence encoding the Homo sapiens(Human) Pulmonary surfactant-associated protein A1, was expressed in the hosts and tags indicated.
469.00 € - 1630.00 €
Specifications
Accession Number | NP_001087239.2 |
---|---|
Gene ID (Entrez) | 653509 |
Name | Pulmonary surfactant-associated protein A1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15932738
|
enQuireBio™
QP10005-IV-100UG |
100 μg |
1630.00 €
100µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15952738
|
enQuireBio™
QP10005-IV-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15942738
|
enQuireBio™
QP10005-IV-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001087239.2 | |
Pulmonary surfactant-associated protein A1 Protein | |
Not determined. | |
Full Length | |
Human | |
His-B2M | |
Tris-based buffer, 50% glycerol |
653509 | |
Research Use Only | |
SFTPA1 | |
Recombinant Protein | |
E. coli | |
EVKDVCVGSP GIPGTPGSHG LPGRDGRDGL KGDPGPPGPM GPPGEMPCPP GNDGLPGAPG IPGECGEKGE PGERGPPGLP AHLDEELQAT LHDFRHQILQ TRGALSLQGS IMTVGEKVFS SNGQSITFDA IQEACARAGG RIAVPRNPEE NEAIASFVKK YNTYAYVGLT EGPSPGDFRY SDGTPVNYTN WYRGEPAGRG KEQCVEMYTD GQWNDRNCLY SRLTICEF | |
Greater than 80% as determined by SDS-PAGE. |