missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Rab GDP dissociation inhibitor beta Protein
A DNA sequence encoding the Homo sapiens (Human) Rab GDP dissociation inhibitor beta, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8339-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001108628.1 | |
Rab GDP dissociation inhibitor beta Protein | |
Research Use Only | |
GDI2 | |
Recombinant Protein | |
E. coli | |
MNEEYDVIVL GTGLTECILS GIMSVNGKKV LHMDRNPYYG GESASITPLE DLYKRFKIPG SPPESMGRGR DWNVDLIPKF LMANGQLVKM LLYTEVTRYL DFKVTEGSFV YKGGKIYKVP STEAEALASS LMGLFEKRRF RKFLVYVANF DEKDPRTFEG IDPKKTTMRD VYKKFDLGQD VIDFTGHALA LYRTDDYLDQ PCYETINRIK LYSESLARYG KSPYLYPLYG LGELPQGFAR LSAIYGGTYM LNKPIEEIIV QNGKVIGVKS EGEIARCKQL ICDPSYVKDR VEKVGQVIRV ICILSHPIKN TNDANSCQII IPQNQVNRKS DIYVCMISFA HNVAAQGKYI AIVSTTVETK EPEKEIRPAL ELLEPIEQKF VSISDLLVPK DLGTESQIFI SRTYDATTHF ETTCDDIKNI YKRMTGSEFD FEEMKRKKND I | |
Greater than 90% as determined by SDS-PAGE. |
2665 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |