missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Regulatory protein E2 Protein
A DNA sequence encoding the Homo sapiens (Human) Regulatory protein E2, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_041328.1 |
---|---|
Gene ID (Entrez) | 1489080 |
Name | Regulatory protein E2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16028720
|
enQuireBio™
QP7408-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16048720
|
enQuireBio™
QP7408-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16038720
|
enQuireBio™
QP7408-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16058720
|
enQuireBio™
QP7408-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16068720
|
enQuireBio™
QP7408-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16078720
|
enQuireBio™
QP7408-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_041328.1 | |
Regulatory protein E2 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
1489080 | |
Research Use Only | |
E2 | |
Recombinant Protein | |
E. coli | |
METLCQRLNV CQDKILTHYE NDSTDLRDHI DYWKHMRLEC AIYYKAREMG FKHINHQVVP TLAVSKNKAL QAIELQLTLE TIYNSQYSNE KWTLQDVSLE VYLTAPTGCI KKHGYTVEVQ FDGDICNTMH YTNWTHIYIC EEASVTVVEG QVDYYGLYYV HEGIRTYFVQ FKDDAEKYSK NKVWEVHAGG QVILCPTSVF SSNEVSSPEI IRQHLANHPA ATHTKAVALG TEETQTTIQR PRSEPDTGNP CHTTKLLHRD SVDSAPILTA FNSSHKGRIN CNSNTTPIVH LKGDANTLKC LRYRFKKHCT LYTAVSSTWH WTGHNVKHKS AIVTLTYDSE WQRDQFLSQV KIPKTITVST GFMSI | |
Greater than 90% as determined by SDS-PAGE. |