missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Rhombotin-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Rhombotin-1, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001257357.1 |
---|---|
Gene ID (Entrez) | 4004 |
Name | Rhombotin-1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16044391
|
enQuireBio™
QP8396-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16064391
|
enQuireBio™
QP8396-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16054391
|
enQuireBio™
QP8396-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16074391
|
enQuireBio™
QP8396-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16084391
|
enQuireBio™
QP8396-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16094391
|
enQuireBio™
QP8396-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001257357.1 | |
Rhombotin-1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
4004 | |
Research Use Only | |
LMO1 | |
Recombinant Protein | |
E. coli | |
DKEDGVPMLS VQPKGKQKGC AGCNRKIKDR YLLKALDKYW HEDCLKCACC DCRLGEVGST LYTKANLILC RRDYLRLFGT TGNCAACSKL IPAFEMVMRA RDNVYHLDCF ACQLCNQRFC VGDKFFLKNN MILCQMDYEE GQLNGTFESQ VQ | |
Greater than 90% as determined by SDS-PAGE. |