missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human S-arrestin Protein
A DNA sequence encoding the Homo sapiens (Human) S-arrestin, was expressed in the hosts and tags indicated.
287.00 € - 3175.00 €
Specifications
Accession Number | NP_000532.2 |
---|---|
Gene ID (Entrez) | 6295 |
Name | S-arrestin Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16048211
|
enQuireBio™
QP9381-YE-100UG |
100 μg |
880.00 €
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16088211
|
enQuireBio™
QP9381-YE-500UG |
500 μg |
2535.00 €
500µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16058211
|
enQuireBio™
QP9381-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16078211
|
enQuireBio™
QP9381-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16098211
|
enQuireBio™
QP9381-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16068211
|
enQuireBio™
QP9381-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000532.2 | |
S-arrestin Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
6295 | |
Research Use Only | |
SAG | |
Recombinant Protein | |
Yeast | |
MAASGKTSKS EPNHVIFKKI SRDKSVTIYL GNRDYIDHVS QVQPVDGVVL VDPDLVKGKK VYVTLTCAFR YGQEDIDVIG LTFRRDLYFS RVQVYPPVGA ASTPTKLQES LLKKLGSNTY PFLLTFPDYL PCSVMLQPAP QDSGKSCGVD FEVKAFATDS TDAEEDKIPK KSSVRLLIRK VQHAPLEMGP QPRAEAAWQF FMSDKPLHLA VSLNKEIYFH GEPIPVTVTV TNNTEKTVKK IKAFVEQVAN VVLYSSDYYV KPVAMEEAQE KVPPNSTLTK TLTLLPLLAN NRERRGIALD GKIKHEDTNL ASSTIIKEGI DRTVLGILVS YQIKVKLTVS GFLGELTSSE VATEVPFRLM HPQPEDPAKE SYQDANLVFE EFARHNLKDA GEAEEGKRDK NDVDE | |
Greater than 90% as determined by SDS-PAGE. |