missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human S100B Protein
A cDNA sequence encoding the S100B was constructed and used to recombinantly synthesize the protein.
264.00 € - 715.00 €
Specifications
Gene ID (Entrez) | 6285 |
---|---|
Name | S100B Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | S100B |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15933459
|
enQuireBio™
QP13402-GST-2UG |
2 μg |
264.00 €
2µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15923459
|
enQuireBio™
QP13402-GST-10UG |
10 μg |
715.00 €
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
6285 | |
Research Use Only | |
S100B | |
Human | |
GST | |
S100B is supplied in 50mM Tris-Acetate, pH 7.5, 1mM EDTA and 20% Glycerol. |
S100B Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MSELEKAMVA LIDVFHQYSG REGDKHKLKK SELKELINNE LSHFLEEIKE QEVVDKVMET LDNDGDGECD FQEFMAFVAM VTTACHEFFE HE |