missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SCF / C-kit ligand Protein

A DNA sequence encoding the Homo sapiens (Human) SCF / C-kit ligand, was expressed in the hosts and tags indicated.

160.00 € - 2285.00 €

Specifications

Accession Number NP_000890.1
Gene ID (Entrez) 4254
Name SCF / C-kit ligand Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16012030
View Documents Promotion Details
enQuireBio™
QP6264-EC-100UG
100 μg
505.00 €
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16032030
View Documents Promotion Details
enQuireBio™
QP6264-EC-1MG
1 mg
2285.00 €
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16022030
View Documents Promotion Details
enQuireBio™
QP6264-EC-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16042030
View Documents Promotion Details
enQuireBio™
QP6264-EC-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16052030
View Documents Promotion Details
enQuireBio™
QP6264-EC-500UG
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16062030
View Documents Promotion Details
enQuireBio™
QP6264-EC-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_000890.1
SCF / C-kit ligand Protein
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
4254
Research Use Only
Kitlg
Recombinant Protein
E. coli
EGICRNRVTN NVKDVTKLVA NLPKDYMITL KYVPGMDVLP SHCWISEMVV QLSDSLTDLL DKFSNISEGL SNYSIIDKLV NIVDDLVECV KENSSKDLKK SFKSPEPRLF TPEEFFRIFN RSIDAFKDFV VASETSDCVV SSTLSPEKDS RVSVTKPFML PPVA
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications