missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SERPINA3 Protein
A cDNA sequence encoding the SERPINA3 was constructed and used to recombinantly synthesize the protein.
131.00 € - 3275.00 €
Specifications
Name | SERPINA3 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15933629
|
enQuireBio™
QP13456-HIS-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15923629
|
enQuireBio™
QP13456-HIS-25UG |
25 μg |
203.00 €
25µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15913629
|
enQuireBio™
QP13456-HIS-1MG |
1 mg |
3275.00 €
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
SERPINA3 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
Lyophilized from 0.02M NH4HCO3. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MERMLPLLAL GLLAAGFCPA VLCHPNSPLD EENLTQENQD RGTHVDLGLA SANVDFAFSL YKQLVLKAPD KNVIFSPLSI STALAFLSLG AHNTTLTEIL KGLKFNLTET SEAEIHQSFQ HLLRTLNQSS DELQLSMGNA MFVKEQLSLL DRFTEDAKRL YGSEAFATDF QDSAAAKKLI NDYVKNGTRG KITDLIKDLD SQTMMVLVNY IFFKAKWEMP FDPQDTHQSR FYLSKKKWVM VPMMSLHHLT IPYFRDEELS CTVVELKYTG NASALFILPD QDKMEEVEAM LLPETLKRWR DSLEFREIGE LYLPKFSISR DYNLNDILLQ LGIEEAFTSK ADLSGITGAR NLAVSQVVHK AVLDVFEEGT EASAATAVKI TLLSALVETR TIVRFNRPFL MIIVPTDTQN IFFMSKVTNP KQA | |
Greater than 99.0%. |