missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SerpinB9 / CAP-3 Protein
A DNA sequence encoding the Homo sapiens (Human) SerpinB9 / CAP-3, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_004146.1 |
---|---|
Gene ID (Entrez) | 5272 |
Name | SerpinB9 / CAP-3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16024980
|
enQuireBio™
QP6671-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16044980
|
enQuireBio™
QP6671-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16034980
|
enQuireBio™
QP6671-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16054980
|
enQuireBio™
QP6671-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16064980
|
enQuireBio™
QP6671-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16074980
|
enQuireBio™
QP6671-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_004146.1 | |
SerpinB9 / CAP-3 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
5272 | |
Research Use Only | |
SerpinB9 | |
Recombinant Protein | |
E. coli | |
METLSNASGT FAIRLLKILC QDNPSHNVFC SPVSISSALA MVLLGAKGNT ATQMAQALSL NTEEDIHRAF QSLLTEVNKA GTQYLLRTAN RLFGEKTCQF LSTFKESCLQ FYHAELKELS FIRAAEESRK HINTWVSKKT EGKIEELLPG SSIDAETRLV LVNAIYFKGK WNEPFDETYT REMPFKINQE EQRPVQMMYQ EATFKLAHVG EVRAQLLELP YARKELSLLV LLPDDGVELS TVEKSLTFEK LTAWTKPDCM KSTEVEVLLP KFKLQEDYDM ESVLRHLGIV DAFQQGKADL SAMSAERDLC LSKFVHKSFV EVNEEGTEAA AASSCFVVAE CCMESGPRFC ADHPFLFFIR HNRANSILFC GRFSSP | |
Greater than 80% as determined by SDS-PAGE. |