missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SERTAD2 Protein
A cDNA sequence encoding the SERTAD2 was constructed and used to recombinantly synthesize the protein.
131.00 € - 1740.00 €
Specifications
Gene ID (Entrez) | 9792 |
---|---|
Name | SERTAD2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | SERTAD2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15983679
|
enQuireBio™
QP13471-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15973679
|
enQuireBio™
QP13471-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15963679
|
enQuireBio™
QP13471-100UG |
100 μg |
1740.00 €
100µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
9792 | |
Research Use Only | |
SERTAD2 | |
Human | |
His | |
The SERTAD2 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |
SERTAD2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMLGKGGK RKFDEHEDGL EGKIVSPCDG PSKVSYTLQR QTIFNISLMK LYNHRPLTEP SLQKTVLINN MLRRIQEELK QEGSLRPMFT PSSQPTTEPS DSYREAPPAF SHLASPSSHP CDLGSTTPLE ACLTPASLLE DDDDTFCTSQ AMQPTAPTKL SPPALLPEKD SFSSALDEIE ELCPTSTSTE AATAATDSVK GTSSEAGTQK LDGPQESRAD DSKLMDSLPG NFEITTSTGF LTDLTLDDIL FADIDTSMYD FDPCTSSSGT ASKMAPVSAD DLLKTLAPYS SQPVTPSQPF KMDLTELDHI MEVLVGS | |
Greater than 90% as determined by SDS-PAGE. |