missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SH2D1A Protein
A cDNA sequence encoding the SH2D1A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13485-1mg
Additional Details : Weight : 0.01000kg
Specifications
SH2D1A Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MDAVAVYHGK ISRETGEKLL LATGLDGSYL LRDSESVPGV YCLCVLYHGY IYTYRVSQTE TGSWSAETAP GVHKRYFRKI KNLISAFQKP DQGIVIPLQY PVEKKSSARS TQGTTGIRED PDVCLKAP | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The SH2D1A solution (1 mg/ml) contains 20mM Tris-HCl Buffer (pH 7.5), 1mM DTT and 10% Glycerol. |