missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SIRP alpha / CD172a Protein
A DNA sequence encoding the Homo sapiens (Human) SIRP alpha / CD172a, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6684-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001035111.1 | |
SIRP alpha / CD172a Protein | |
Research Use Only | |
SIRPA | |
Recombinant Protein | |
E. coli | |
EEELQVIQPD KSVLVAAGET ATLRCTATSL IPVGPIQWFR GAGPGRELIY NQKEGHFPRV TTVSDLTKRN NMDFSIRIGN ITPADAGTYY CVKFRKGSPD DVEFKSGAGT ELSVRAKPSA PVVSGPAARA TPQHTVSFTC ESHGFSPRDI TLKWFKNGNE LSDFQTNVDP VGESVSYSIH STAKVVLTRE DVHSQVICEV AHVTLQGDPL RGTANLSETI RVPPTLEVTQ QPVRAENQVN VTCQVRKFYP QRLQLTWLEN GNVSRTETAS TVTENKDGTY NWMSWLLVNV SAHRDDVKLT CQVEHDGQPA VSKSHDLKVS AHPKEQGSNT AAENTGSNER NIY | |
Greater than 90% as determined by SDS-PAGE. |
140885 | |
500 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |