missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SIX1 Protein
A cDNA sequence encoding the SIX1 was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Name | SIX1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15903819
|
enQuireBio™
QP13513-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15993809
|
enQuireBio™
QP13513-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15983809
|
enQuireBio™
QP13513-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
SIX1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The SIX1 solution contains 20mM Tris-HCl buffer (pH 8.0), 2M Urea and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMSMLPS FGFTQEQVAC VCEVLQQGGN LERLGRFLWS LPACDHLHKN ESVLKAKAVV AFHRGNFREL YKILESHQFS PHNHPKLQQL WLKAHYVEAE KLRGRPLGAV GKYRVRRKFP LPRTIWDGEE TSYCFKEKSR GVLREWYAHN PYPSPREKRE LAEATGLTTT QVSNWFKNRR QRDRAAEAKE RENTENNNSS SNKQNQLSPL EGGKPLMSSS EEEFSPPQSP DQNSVLLLQG NMGHARSSNY SLPGLTASQP SHGLQTHQHQ LQDSLLGPLT SSLVDLGS | |
Greater than 85% as determined by SDS-PAGE. |