missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SNAPIN Protein

A cDNA sequence encoding the SNAPIN was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13534-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15983879

  • 2610.00 € / 1mg
Estimated Shipment: 24-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

SNAPIN Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MAGAGSAAVS GAGTPVAGPT GRDLFAEGLL EFLRPAVQQL DSHVHAVRES QVELREQIDN LATELCRINEDQKVALDLDP YVKKLLNARR RVVLVNNILQ NAQERLRRLN HSVAKETARR RAMLDSGIYP PGSPGK
Greater than 90.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The protein solution contains 20mM Tris-HCl pH 8.0, 5mM DTT, 2mM EDTA, 0.2M NaCl, and 40% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers