missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SOCS3 Protein

A cDNA sequence encoding the SOCS3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13557-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15983959

Quantity Price
1 171.64 € / 10µg
10 161.27 € / 10µg
Estimated Shipment: 26-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

SOCS3 Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
SOCS3 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol.
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSHMVT HSKFPAAGMS RPLDTSLRLK TFSSKSEYQL VVNAVRKLQE SGFYWSAVTG GEANLLLSAE PAGTFLIRDS SDQRHFFTLS VKTQSGTKNL RIQCEGGSFS LQSDPRSTQP VPRFDCVLKL VHHYMPPPGA PSFPSPPTEP SSEVPEQPSA QPLPGSPPRR AYYIYSGGEK IPLVLSRPLS SNVATLQHLC RKTVNGHLDS YEKVTQLPGP IREFLDQYDA PL
Videos
SDS
Documents
Certificates