missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SOST Protein
A cDNA sequence encoding the SOST was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13561-HIS-HEK-10ug
Additional Details : Weight : 0.01000kg
Specifications
SOST Protein | |
Research Use Only | |
Recombinant Protein | |
Human (HEK293) | |
MKHHHHHHASQGWQAFKNDA TEIIPELGEY PEPPPELENN KTMNRAENGG RPPHHPFETK DVSEYSCREL HFTRYVTDGP CRSAKPVTEL VCSGQCGPAR LLPNAIGRGK WWRPSGPDFR CIPDRYRAQR VQLLCPGGEA PRARKVRLVA SCKCKRLTRF HNQSELKDFG TEAARPQKGR KPRPRARSAK ANQAELENAY | |
Greater than 90.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SOST filtered (0.4 microm) and lyophilized from 0.5 mg/ml in 0.03M Acetate buffer pH 4.0. |