missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SOST Protein
A cDNA sequence encoding the SOST was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Name | SOST Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15953979
|
enQuireBio™
QP13561-HIS-HEK-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15933979
|
enQuireBio™
QP13561-HIS-HEK-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15943979
|
enQuireBio™
QP13561-HIS-HEK-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
SOST Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SOST filtered (0.4 microm) and lyophilized from 0.5 mg/ml in 0.03M Acetate buffer pH 4.0. |
Research Use Only | |
Recombinant Protein | |
Human (HEK293) | |
MKHHHHHHASQGWQAFKNDA TEIIPELGEY PEPPPELENN KTMNRAENGG RPPHHPFETK DVSEYSCREL HFTRYVTDGP CRSAKPVTEL VCSGQCGPAR LLPNAIGRGK WWRPSGPDFR CIPDRYRAQR VQLLCPGGEA PRARKVRLVA SCKCKRLTRF HNQSELKDFG TEAARPQKGR KPRPRARSAK ANQAELENAY | |
Greater than 90.0% as determined by SDS-PAGE. |