missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Sperm surface protein Sp17 Protein
A DNA sequence encoding the Homo sapiens (Human) Sperm surface protein Sp17, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8737-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_059121.1 | |
Sperm surface protein Sp17 Protein | |
Research Use Only | |
SPA17 | |
Recombinant Protein | |
E. coli | |
MSIPFSNTHY RIPQGFGNLL EGLTREILRE QPDNIPAFAA AYFESLLEKR EKTNFDPAEW GSKVEDRFYN NHAFEEQEPP EKSDPKQEES QISGKEEETS VTILDSSEED KEKEEVAAVK IQAAFRGHIA REEAKKMKTN SLQNEE | |
Greater than 90% as determined by SDS-PAGE. |
53340 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |