missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SPP1 Protein
A cDNA sequence encoding the SPP1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13580-HIS-HEK-1mg
Additional Details : Weight : 0.01000kg
Specifications
SPP1 Protein | |
Research Use Only | |
Recombinant Protein | |
Human (HEK293) | |
IPVKQADSGSSEEKQLYNKYPDAVATWLNPDPSQKQNLLAPQNAVSSEETNDFKQETLPSKSNESHDHMDDMDDEDDDDHVDSQDSIDSNDSDDVDDTDDSHQSDESHHSDESDELVTDFPTDLPATEVFTPVVPTVDTYDGRGDSVVYGLRSKSKKFRRPDIQYPDATDEDITSHMESEELNGAYKAIPVAQDLNAPSDWDSRGKDSYETSQLDDQSAETHSHKQSRLYKRKANDESNEHSDVIDSQELSKVSREFHSHEFHSHEDMLVVDPKSKEEDKHLKFRISHELDSASSEVNVDHHHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
Osteopontin was lyophilized from a 0.2microm filtered solution of 20mM PB and 150mM NaCl, pH 7.2. |