missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SPRR1B Protein
A cDNA sequence encoding the SPRR1B was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13584-10ug
Additional Details : Weight : 0.01000kg
Specifications
SPRR1B Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHAS SQQQKQPCTP PPQLQQQQVK QPCQPPPQEP CIPKTKEPCH PKVPEPCHPK VPEPCQPKVP EPCHPKVPEP CPSIVTPAPA QQKTKQK | |
Greater than 95.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SPRR1B filtered (0.4 microm) and lyophilized from 0.5 mg/ml in 0.05M phosphate buffer and 0.075M NaCl, pH 7.4. |