missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ST3GAL5 Protein
A cDNA sequence encoding the ST3GAL5 was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Name | ST3GAL5 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15974109
|
enQuireBio™
QP13602-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15964109
|
enQuireBio™
QP13602-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15954109
|
enQuireBio™
QP13602-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
ST3GAL5 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
ST3GAL5 protein solution (1 mg/ml) containing In 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSLKLNYTT EECDMKKMHY VDPDHVKRAQ KYAQQVLQKE CRPKFAKTSM ALLFEHRYSV DLLPFVQKAP KDSEAESKYD PPFGFRKFSS KVQTLLELLP EHDLPEHLKA KTCRRCVVIG SGGILHGLEL GHTLNQFDVV IRLNSAPVEG YSEHVGNKTT IRMTYPEGAP LSDLEYYSND LFVAVLFKSV DFNWLQAMVK KETLPFWVRL FFWKQVAEKI PLQPKHFRIL NPVIIKETAF DILQYSEPQS RFWGRDKNVP TIGVIAVVLA THLCDEVSLA GFGYDLNQPR TPLHYFDSQC MAAMNFQTMH NVTTETKFLL KLVKEGVVKD LSGGIDREF | |
Greater than 85.0% as determined by SDS-PAGE. |