missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human STOML1 Protein
A cDNA sequence encoding the STOML1 was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Name | STOML1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15934179
|
enQuireBio™
QP13623-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15914179
|
enQuireBio™
QP13623-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15924179
|
enQuireBio™
QP13623-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
STOML1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
STOML1 protein solution (0.5 mg/ml) containing 20mM Tris-HCl (pH 8.0) and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSLKIVPTY ERMIVFRLGR IRTPQGPGMV LLLPFIDSFQ RVDLRTRAFN VPPCKLASKD GAVLSVGADV QFRIWDPVLS VMTVKDLNTA TRMTAQNAMT KALLKRPLRE IQMEKLKISD QLLLEINDVT RAWGLEVDRV ELAVEAVLQP PQDSPAGPNL DSTLQQLALH FLGGSMNSMA GGAPSPGPAD TVEMVSEVEP PAPQVGARSS PKQPLAEGLL TALQPFLSEA LVSQVGACYQ FNVVLPSGTQ SAYFLDLTTG RGRVGHGVPD GIPDVVVEMA EADLRALLCR ELRPLGAYMS GRLKVKGDLA MAMKLEAVLR ALK | |
Greater than 90.0% as determined by SDS-PAGE. |