missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SUGT1 Protein
A cDNA sequence encoding the SUGT1 was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Name | SUGT1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15934239
|
enQuireBio™
QP13640-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15924239
|
enQuireBio™
QP13640-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15914239
|
enQuireBio™
QP13640-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
SUGT1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The SGT1 protein solution (0.5 mg/1ml) is formulated in 20 mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 1mM DTT and 20% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MHRVGQAGLQ LLTSSDPPAL DSQSAGITGA DANFSVWIKR CQEAQNGSES EVWTHQSKIK YDWYQTESQVVITLMIKNVQ KNDVNVEFSE KELSALVKLP SGEDYNLKLE LLHPIIPEQS TFKVLSTKIE IKLKKPEAVR WEKLEGQGDV PTPKQFVADVKNLYPSSSPY TRNWDKLVGE IKEEEKNEKL EGDAALNRLF QQIYSDGSDE VKRAMNKSFM ESGGTVLSTN WSDVGKRKVE INPPDDMEWKKY | |
Greater than 90% as determined by SDS-PAGE. |