missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SURF1 Protein

A cDNA sequence encoding the SURF1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13651-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15984269

Quantity Price
1 2841.43 € / 1mg
3 2672.08 € / 1mg
Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

SURF1 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The Surfeit-1 solution contains 20 mM Tris-HCl buffer (pH 8.0), 0.1M Nacl, 1mM DTT and 20% glycerol.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MQVQRRKWKL NLIAELESRV LAEPVPLPAD PMELKNLEYR PVKVRGCFDH SKELYMMPRT MVDPVREARE GGLISSSTQS GAYVVTPFHC TDLGVTILVN RGFVPRKKVN PETRQKGQIE GEVDLIGMVR LTETRQPFVP ENNPERNHWH YRDLEAMARI TGAEPIFIDA NFQSTVPGGP IGGQTRVTLR NEHLQ
Videos
SDS
Documents
Certificates