missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SYCE3 Protein
A cDNA sequence encoding the SYCE3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13654-10ug
Additional Details : Weight : 0.01000kg
Specifications
644186 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MDDADPEERN YDNMLKMLSD LNKDLEKLLE EMEKISVQAT WMAYDMVVMR TNPTLAESMR RLEDAFVNCK EEMEKNWQEL LHETKQRL | |
Greater than 90% as determined by SDS-PAGE. |
SYCE3 Protein | |
Research Use Only | |
SYCE3 | |
Human | |
His | |
The SYCE3 solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 200mM NaCl, 2mM DTT and 20% glycerol. |