missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human T-box transcription factor TBX18 Protein
A DNA sequence encoding the Homo sapiens (Human) T-box transcription factor TBX18, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001073977.1 |
---|---|
Gene ID (Entrez) | 9096 |
Name | T-box transcription factor TBX18 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16015680
|
enQuireBio™
QP6766-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16035680
|
enQuireBio™
QP6766-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16025680
|
enQuireBio™
QP6766-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16045680
|
enQuireBio™
QP6766-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16055680
|
enQuireBio™
QP6766-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16065680
|
enQuireBio™
QP6766-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001073977.1 | |
T-box transcription factor TBX18 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
9096 | |
Research Use Only | |
TBX18 | |
Recombinant Protein | |
E. coli | |
MAEKRRGSPC SMLSLKAHAF SVEALIGAEK QQQLQKKRRK LGAEEAAGAV DDGGCSRGGG AGEKGSSEGD EGAALPPPAG ATSGPARSGA DLERGAAGGC EDGFQQGASP LASPGGSPKG SPARSLARPG TPLPSPQAPR VDLQGAELWK RFHEIGTEMI ITKAGRRMFP AMRVKISGLD PHQQYYIAMD IVPVDNKRYR YVYHSSKWMV AGNADSPVPP RVYIHPDSPA SGETWMRQVI SFDKLKLTNN ELDDQGHIIL HSMHKYQPRV HVIRKDCGDD LSPIKPVPSG EGVKAFSFPE TVFTTVTAYQ NQQITRLKID RNPFAKGFRD SGRNRMGLEA LVESYAFWRP SLRTLTFEDI PGIPKQGNAS SSTLLQGTGN GVPATHPHLL SGSSCSSPAF HLGPNTSQLC SLAPADYSAC ARSGLTLNRY STSLAETYNR LTNQAGETFA PPRTPSYVGV SSSTSVNMSM GGTDGDTFSC PQTSLSMQIS GMSPQLQYIM PSPSSNAFAT NQTHQGSYNT FRLHSPCALY GYNFSTSPKL AASPEKIVSS QGSFLGSSPS GTMTDRQMLP PVEGVHLLSS GGQQSFFDSR TLGSLTLSSS QVSAHMV | |
Greater than 90% as determined by SDS-PAGE. |