missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TACI Protein

A cDNA sequence encoding the TACI was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13681-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15984359

  • 3401.86 € / 1mg

Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

23495
Greater than 90% as determined by SDS-PAGE.
Research Use Only
TACI
Human
His
The TACI solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea.
TACI Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMSGLGRS RRGGRSRVDQ EERFPQGLWT GVAMRSCPEE QYWDPLLGTC MSCKTICNHQ SQRTCAAFCR SLSCRKEQGK FYDHLLRDCI SCASICGQHP KQCAYFCENK LRSPVNLPPE LRRQRSGEVE NNSDNSGRYQ GLEHRGSEAS PALPGLKLSA DQVALVYS
Videos
SDS
Documents
Certificates