missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TACI Protein
A cDNA sequence encoding the TACI was constructed and used to recombinantly synthesize the protein.
131.00 € - 6790.00 €
Specifications
Gene ID (Entrez) | 23495 |
---|---|
Name | TACI Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | TACI |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15985428
|
enQuireBio™
QP10873-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15975428
|
enQuireBio™
QP10873-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965428
|
enQuireBio™
QP10873-1MG |
1 mg |
6790.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
23495 | |
Research Use Only | |
TACI | |
Recombinant Protein | |
E. coli | |
MSGLGRSRRG GRSRVDQEER FPQGLWTGVA MRSCPEEQYW DPLLGTCMSC KTICNHQSQR TCAAFCRSLS CRKEQGKFYD HLLRDCISCA SICGQHPKQC AYFCENKLRS PVNLPPELRR QRSGEVENNS DNSGRYQGLE HRGSEASPAL PGLKLSADQV | |
Greater than 95.0% as determined by: (a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
TACI Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Fully biologically active when compared to standard. The biological activity is determined by its ability to block human BAFF induced T2B cell survival using a concentration range of 1.0-3.0 microg/ml corresponding to a specific activity of 334-1000 IU/mg. | |
Human | |
Untagged | |
TACI protein was lyophilized from a 0.2 microm filtered concentrated solution in PBS, pH 7.4. |