missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TCEAL1 Protein

A cDNA sequence encoding the TCEAL1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13706-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15984439

Quantity Price
1 114.77 € / 5µg
10 107.33 € / 5µg
Estimated Shipment: 23-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

TCEAL1 Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The TCEAL1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 2mM DTT, 20% glycerol and 0.1M NaCl.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MDKPRKENEE EPQSAPKTDE ERPPVEHSPE KQSPEEQSSE EQSSEEEFFP EELLPELLPE MLLSEERPPQ EGLSRKDLFE GRPPMEQPPC GVGKHKLEEG SFKERLARSR PQFRGDIHGR NLSNEEMIQA ADELEEMKRV RNKLMIMHWK AKRSRPYPIL EHHHHHH
Videos
SDS
Documents
Certificates