missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TCEB2 Protein
A DNA sequence encoding the Homo sapiens (Human) TCEB2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7575-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_009039.1 | |
TCEB2 Protein | |
Research Use Only | |
TCEB2 | |
Recombinant Protein | |
E. coli | |
MDVFLMIRRH KTTIFTDAKE SSTVFELKRI VEGILKRPPD EQRLYKDDQL LDDGKTLGEC GFTSQTARPQ APATVGLAFR ADDTFEALCI EPFSSPPELP DVMKPQDSGS SANEQAVQ | |
Greater than 80% as determined by SDS-PAGE. |
6923 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |