missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TCL1A Protein
A cDNA sequence encoding the TCL1A was constructed and used to recombinantly synthesize the protein.
131.00 € - 2610.00 €
Specifications
Name | TCL1A Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15944459
|
enQuireBio™
QP13711-10UG |
10 μg |
131.00 €
10µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15964459
|
enQuireBio™
QP13711-50UG |
50 μg |
203.00 €
50µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15954459
|
enQuireBio™
QP13711-1MG |
1 mg |
2610.00 €
1mg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
TCL1A Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The TCL1A protein solution contains 50mM Tris-HCl buffer (pH 7.5) and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MAECPTLGEA VTDHPDRLWA WEKFVYLDEK QHAWLPLTIE IKDRLQLRVL LRREDVVLGR PMTPTQIGPS LLPIMWQLYP DGRYRSSDSS FWRLVYHIKI DGVEDMLLEL LPDD | |
Greater than 90.0% as determined by SDS-PAGE. |