missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Tegument protein VP16 Protein
A DNA sequence encoding the Homo sapiens (Human) Tegument protein VP16, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Name | Tegument protein VP16 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | UL48 |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16037990
|
enQuireBio™
QP7282-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16057990
|
enQuireBio™
QP7282-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16047990
|
enQuireBio™
QP7282-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16067990
|
enQuireBio™
QP7282-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16077990
|
enQuireBio™
QP7282-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16087990
|
enQuireBio™
QP7282-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Tegument protein VP16 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
UL48 | |
Recombinant Protein | |
E. coli | |
MDLLVDELFA DMDADGASPP PPRPAGGPKN TPAAPPLYAT GRLSQAQLMP SPPMPVPPAA LFNRLLDDLG FSAGPALCTM LDTWNEDLFS ALPTNADLYR ECKFLSTLPS DVVEWGDAYV PERAQIDIRA HGDVAFPTLP ATRDGLGLYY EALSRFFHAE LRAREESYRT VLANFCSALY RYLRASVRQL HRQAHMRGRD RDLGEMLRAT IADRYYRETA RLARVLFLHL YLFLTREILW AAYAEQMMRP DLFDCLCCDL ESWRQLAGLF QPFMFVNGAL TVRGVPIEAR RLRELNHIRE HLNLPLVRSA ATEEPGAPLT TPPTLHGNQA RASGYFMVLI RAKLDSYSSF TTSPSEAVMR EHAYSRARTK NNYGSTIEGL LDLPDDDAPE EAGLAAPRLS FLPAGHTRRL STAPPTDVSL GDELHLDGED VAMAHADALD DFDLDMLGDG DSPGPGFTPH DSAPYGALDM ADFEFEQMFT DALGIDEYGG | |
Greater than 90% as determined by SDS-PAGE. |