missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TEK Protein
A cDNA sequence encoding the TEK was constructed and used to recombinantly synthesize the protein.
131.00 € - 5510.00 €
Specifications
Name | TEK Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15904489
|
enQuireBio™
QP13718-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15984479
|
enQuireBio™
QP13718-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15994479
|
enQuireBio™
QP13718-1MG |
1 mg |
5510.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
TEK Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
TEK Fc Chimera was lyophilized from a concentrated (1 mg/ml) sterile solution containing 1xPBS. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
MDSLASLVLC GVSLLLSGTV EGAMDLILIN SLPLVSDAET SLTCIASGWR PHEPITIGRD FEALMNQHQD PLEVTQDVTR EWAKKVVWKR EKASKINGAY FCEGRVRGEA IRIRTMKMRQ QASFLPATLT MTVDKGDNVN ISFKKVLIKE EDAVIYKNGS FIHSVPRHEV PDILEVHLPH AQPQDAGVYS ARYIGGNLFT SAFTRLIVRR CEAQKWGPEC NHLCTACMNN GVCHEDTGEC ICPPGFMGRT CEKACELHTF GRTCKERCSG QEGCKSYVFC LPDPYGCSCA TGWKGLQCNE ACHPGFYGPD CKLRCSCNNG EMCDRFQGCL CSPGWQGLQC EREGIQRMTP KIVDLPDHIE VNSGKFNPIC KASGWPLPTN EEMTLVKPDG TVLHPKDFNH TDHFSVAIFT IHRILPPDSG VWVCSVNTVA GMVEKPFNIS VKVLPKPLNA PNVIDTGHNF AVINISSEPY FGDGPIKSKK LLYKPVNHYE AWQHIQVTNE IVTLNYLEPR TEYELCVQLV RRGEGGEGHP GPVRRFTTAS IGLPPPRGLN LLPKSQTTLN LTWQPIFPSS EDDFYVEVER RSVQKSDQQN IKVPGNLTSV LLNNLHPREQ YVVRARVNTK AQGEWSEDLT AWTLSDILPP QPENIKISNI THSSAVISWT ILDGYSISSI TIRYKVQGKN EDQHVDVKIK NATITQYQLK GLEPETAYQV DIFAENNIGS SNPAFSHELV TLPESQAPAD LGGGKMLLIA ILGSAGMTCL TVLLAFLIIL QLKRANVQRR MAQAFQNVRE EPAVQFNSGT LALNRKVKNN PDPTIYPVLD WNDIKFQDVI GEGNFGQVLK ARIKKDGLRM DAAIKRMKEY ASKDDHRDFA GELEVLCKLG HHPNIINLLG ACEHRGYLYL AIEYAPHGNL | |
Greater than 90.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |