missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TEK Protein

A cDNA sequence encoding the TEK was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13718-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15984479

Quantity Price
1 171.64 € / 10µg
10 161.27 € / 10µg
Estimated Shipment: 23-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

TEK Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
TEK Fc Chimera was lyophilized from a concentrated (1 mg/ml) sterile solution containing 1xPBS.
Greater than 90.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
Recombinant Protein
Insect
MDSLASLVLC GVSLLLSGTV EGAMDLILIN SLPLVSDAET SLTCIASGWR PHEPITIGRD FEALMNQHQD PLEVTQDVTR EWAKKVVWKR EKASKINGAY FCEGRVRGEA IRIRTMKMRQ QASFLPATLT MTVDKGDNVN ISFKKVLIKE EDAVIYKNGS FIHSVPRHEV PDILEVHLPH AQPQDAGVYS ARYIGGNLFT SAFTRLIVRR CEAQKWGPEC NHLCTACMNN GVCHEDTGEC ICPPGFMGRT CEKACELHTF GRTCKERCSG QEGCKSYVFC LPDPYGCSCA TGWKGLQCNE ACHPGFYGPD CKLRCSCNNG EMCDRFQGCL CSPGWQGLQC EREGIQRMTP KIVDLPDHIE VNSGKFNPIC KASGWPLPTN EEMTLVKPDG TVLHPKDFNH TDHFSVAIFT IHRILPPDSG VWVCSVNTVA GMVEKPFNIS VKVLPKPLNA PNVIDTGHNF AVINISSEPY FGDGPIKSKK LLYKPVNHYE AWQHIQVTNE IVTLNYLEPR TEYELCVQLV RRGEGGEGHP GPVRRFTTAS IGLPPPRGLN LLPKSQTTLN LTWQPIFPSS EDDFYVEVER RSVQKSDQQN IKVPGNLTSV LLNNLHPREQ YVVRARVNTK AQGEWSEDLT AWTLSDILPP QPENIKISNI THSSAVISWT ILDGYSISSI TIRYKVQGKN EDQHVDVKIK NATITQYQLK GLEPETAYQV DIFAENNIGS SNPAFSHELV TLPESQAPAD LGGGKMLLIA ILGSAGMTCL TVLLAFLIIL QLKRANVQRR MAQAFQNVRE EPAVQFNSGT LALNRKVKNN PDPTIYPVLD WNDIKFQDVI GEGNFGQVLK ARIKKDGLRM DAAIKRMKEY ASKDDHRDFA GELEVLCKLG HHPNIINLLG ACEHRGYLYL AIEYAPHGNL LDFLRKSRVL ETDPAFAIAN STASTLSSQQ LLHFAADVAR GMDYLSQKQF IHRDLAARNI LVGENYVAKI ADFGLSRGQE VYVKKTMGRL PVRWMAIESL NYSVYTTNSD VWSYGVLLWE IVSLGGTPYC GMTCAELYEK LPQGYRLEKP LNCDDEVYDL MRQCWREKPY ERPSFAQILV SLNRMLEERK TYVNTTLYEK FTYAGIDCSA EEAA
Videos
SDS
Documents
Certificates