missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Telomerase protein component 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Telomerase protein component 1, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Spécification
Accession Number | NP_009041.2 |
---|---|
Gene ID (Entrez) | 7011 |
Name | Telomerase protein component 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
16037171
|
enQuireBio™
QP8918-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16057171
|
enQuireBio™
QP8918-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16047171
|
enQuireBio™
QP8918-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16067171
|
enQuireBio™
QP8918-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16077171
|
enQuireBio™
QP8918-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16087171
|
enQuireBio™
QP8918-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Spécification
NP_009041.2 | |
Telomerase protein component 1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
7011 | |
Research Use Only | |
TEP1 | |
Recombinant Protein | |
E. coli | |
LKLLCMKPGD APSEIWSSYT ENPMILSTHK EYGIFVLQPK DPGVLSFLRQ KESGEFEERL NFDINLENPS RTLISITQAK PESESSFLCA SSDGILWNLA KCSPEGEWTT GNMWQKKANT PETQTPGTDP STCRESDASM DSDASMDSEP TPHLKTRQRR KIHSGSVTAL HVLPELLVTA SKDRDVKLWE RPSMQLLGLF RCE | |
Greater than 90% as determined by SDS-PAGE. |