missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TES Protein
A cDNA sequence encoding the TES was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13724-2ug
Additional Details : Weight : 0.01000kg
Specifications
TES Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MDLENKVKKM GLGHEQGFGA PCLKCKEKCE GFELHFWRKI CRNCKCGQEE HDVLLSNEED RKVGKLFEDT KYTTLIAKLK SDGIPMYKRN VMILTNPVAA KKNVSINTVT YEWAPPVQNQ ALARQYMQML PKEKQPVAGS EGAQYRKKQL AKQLPAHDQD PSKCHELSPR EVKEMEQFVK KYKSEALGVG DVKLPCEMDA QGPKQMNIPG GDRSTPAAVG AMEDKSAEHK RTQYSCYCCK LSMKEGDPAI YAERAGYDKL WHPACFVCST CHELLVDMIY FWKNEKLYCG RHYCDSEKPR CAGCDELIFS NEYTQAENQN WHLKHFCCFD CDSILAGEIY VMVNDKPVCK PCYVKNHAVV CQGCHNAIDP EVQRVTYNNF SWHASTECFL CSCCSKCLIG QKFMPVEGMV FCSVECKKRM S | |
Greater than 90.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
TES 0.5 mg/ml protein solution contains 20mM Tris pH 8.0, 1mM DTT, 0.1M NaCl and 20% glycerol. |