missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TGFBR2 Protein
A cDNA sequence encoding the TGFBR2 was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Name | TGFBR2 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | In ELISA assay, concentrations 0.5-1microg/ml can be used for TGF-beta measurements. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15905448
|
enQuireBio™
QP10877-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15985438
|
enQuireBio™
QP10877-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15995438
|
enQuireBio™
QP10877-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
TGFBR2 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
HVQK SVNNDMIVTD NNGAVKFPQL CKFCDVRFST CDNQKSCMSN CSITSICEKP QEVCVAVWRK NDENITLETV CHDPKLPYHD FILEDAASPK CIMKEKKKPG ETFFMCSCSS DECNDNIIFS EEYNTSNPD | |
Greater than 97.0% as determined by SDS-PAGE. |
Research Use Only | |
In ELISA assay, concentrations 0.5-1microg/ml can be used for TGF-beta measurements. | |
Human | |
Untagged | |
Lyophilized from a 0.2 ?m filtered solution in buffer containing 150mM NaCl and 50mM sodium phosphate buffer pH 7.5. |