missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TGFBR2 Protein

A cDNA sequence encoding the TGFBR2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10877-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15985438

Quantity Price
1 171.64 € / 10µg
10 161.27 € / 10µg
Estimated Shipment: 26-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

TGFBR2 Protein
10 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
HVQK SVNNDMIVTD NNGAVKFPQL CKFCDVRFST CDNQKSCMSN CSITSICEKP QEVCVAVWRK NDENITLETV CHDPKLPYHD FILEDAASPK CIMKEKKKPG ETFFMCSCSS DECNDNIIFS EEYNTSNPD
Greater than 97.0% as determined by SDS-PAGE.
Research Use Only
In ELISA assay, concentrations 0.5-1microg/ml can be used for TGF-beta measurements.
Human
Untagged
Lyophilized from a 0.2 ?m filtered solution in buffer containing 150mM NaCl and 50mM sodium phosphate buffer pH 7.5.
Videos
SDS
Documents
Certificates