missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human THAP1 Protein
A cDNA sequence encoding the THAP1 was constructed and used to recombinantly synthesize the protein.
131.00 € - 5220.00 €
Specifications
Name | THAP1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15934549
|
enQuireBio™
QP13739-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15924549
|
enQuireBio™
QP13739-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15914549
|
enQuireBio™
QP13739-1MG |
1 mg |
5220.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
THAP1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
THAP1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 2M Urea, 10% glycerol, 1mM DTT and 0.2M NaCl. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMVQSCS AYGCKNRYDK DKPVSFHKFP LTRPSLCKEW EAAVRRKNFK PTKYSSICSE HFTPDCFKRE CNNKLLKENA VPTIFLCTEP HDKKEDLLEP QEQLPPPPLP PPVSQVDAAI GLLMPPLQTP VNLSVFCDHN YTVEDTMHQR KRIHQLEQQV EKLRKKLKTA QQRCRRQERQ LEKLKEVVHF QKEKDDVSER GYVILPNDYF EIVEVPA | |
Greater than 85.0% as determined by SDS-PAGE. |