missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human THAP1 Protein
A cDNA sequence encoding the THAP1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13739-5ug
Additional Details : Weight : 0.01000kg
Specifications
THAP1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMVQSCS AYGCKNRYDK DKPVSFHKFP LTRPSLCKEW EAAVRRKNFK PTKYSSICSE HFTPDCFKRE CNNKLLKENA VPTIFLCTEP HDKKEDLLEP QEQLPPPPLP PPVSQVDAAI GLLMPPLQTP VNLSVFCDHN YTVEDTMHQR KRIHQLEQQV EKLRKKLKTA QQRCRRQERQ LEKLKEVVHF QKEKDDVSER GYVILPNDYF EIVEVPA | |
Greater than 85.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
THAP1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 2M Urea, 10% glycerol, 1mM DTT and 0.2M NaCl. |