missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human THAP7 Protein

A cDNA sequence encoding the THAP7 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13741-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15984549

Quantity Price
1 171.64 € / 20µg
10 161.27 € / 20µg
Estimated Shipment: 23-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

THAP7 Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
THAP7 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea.
Greater than 85.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMPRHCSA AGCCTRDTRE TRNRGISFHR LPKKDNPRRG LWLANCQRLD PSGQGLWDPA SEYIYFCSKH FEEDCFELVG ISGYHRLKEG AVPTIFESFS KLRRTTKTKG HSYPPGPAEV SRLRRCRKRC SEGRGPTTPF SPPPPADVTC FPVEEASAPA TLPASPAGRL EPGLSSPFSD LLGPLGAQAD EAGCSAQPSP ERQPSPLEPR PVSPSAYMLR LPPPAGAYIQ NEHSYQVGSA LLWKRRAEAA LDALDKAQRQ LQACKRREQR LRLRLTKLQQ ERAREKRAQA DARQTLKEHV QDFAMQLSSS MA
Videos
SDS
Documents
Certificates