missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Thrombopoietin / THPO Protein
Recombinant Protein
Brand: enQuireBio™ QP5227-2ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000451.1 | |
Thrombopoietin / THPO Protein | |
2 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 5 ng/mL in the MO7e cell proliferaiton assay. The bioactivity acceptance criteria is set at 2 x 10^5 units/mg. | |
Human | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |
7066 | |
Greater than 90 as determined by SDS-PAGE. | |
Research Use Only | |
THPO | |
Recombinant Protein | |
E. coli | |
MSPAPPACDL RVLSKLLRDS HVLHSRLSQC PEVHPLPTPV LLPAVDFSLG EWKTQMEETK AQDILGAVTL LLEGVMAARG QLGPTCLSSL LGQLSGQVRL LLGALQSLLG TQLPPQGRTT AHKDPNAIFL SFQHLLRGKV RFLMLVGGST LCVRRAPPTT AVPSRTSLVL TLNEL |