missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TIMP1 Protein
A cDNA sequence encoding the TIMP1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13753-HIS-INSECT-2ug
Additional Details : Weight : 0.01000kg
Specifications
TIMP1 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
MAPFEPLASG ILLLLWLIAP SRACTCVPPH PQTAFCNSDL VIRAKFVGTP EVNQTTLYQR YEIKMTKMYK GFQALGDAAD IRFVYTPAME SVCGYFHRSH NRSEEFLIAG KLQDGLLHIT TCSFVAPWNS LSLAQRRGFT KTYTVGCEEC TVFPCLSIPC KLQSGTHCLW TDQLLQGSEK GFQSRHLACL PREPGLCTWQ SLRSQIA | |
Greater than 95.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
TIMP1 protein is supplied in 25mM Sodium acetate pH4.8 and 50% glycerol. |