missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TNC Protein

A cDNA sequence encoding the TNC was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13766-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15984629

Quantity Price
1 171.64 € / 100µg
10 161.27 € / 100µg
Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

TNC Protein
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
The protein solution contains 150mM NaCl, 10mM sodium phosphate, 0.05% NaN3, pH 7.0.
Greater than 95.0% as determined by both:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MDDIYKAAVE QLTEEQKNEF KAAFDIFVLG AEDGCISTKE LGKVMRMLGQ NPTPEELQEM IDEVDEDGSG TVDFDEFLVM MVRCMKDDSK GKSEEELSDL FRMFDKNADG YIDLDELKIM LQATGETITE DDIEELMKDG DKNNDGRIDY DEFLEFMKGV E
Videos
SDS
Documents
Certificates