missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TNFAIP8 Protein
A DNA sequence encoding the Homo sapiens (Human) TNFAIP8, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8735-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001071122.1 | |
TNFAIP8 Protein | |
Research Use Only | |
TNFAIP8 | |
Recombinant Protein | |
E. coli | |
HSEAEESKEV ATDVFNSKNL AVQAQKKILG KMVSKSIATT LIDDTSSEVL DELYRVTREY TQNKKEAEKI IKNLIKTVIK LAILYRNNQF NQDELALMEK FKKKVHQLAM TVVSFHQVDY TFDRNVLSRL LNECREMLHQ IIQRHLTAKS HGRVNNVFDH FSDCEFLAAL YNPFGNFKPH LQKLCDGINK MLDEENI | |
Greater than 90% as determined by SDS-PAGE. |
25816 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |