missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TNFR1 / CD120a / TNFRSF1A Protein
A DNA sequence encoding the Homo sapiens (Human) TNFR1 / CD120a / TNFRSF1A, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001056.1 |
---|---|
Gene ID (Entrez) | 7132 |
Name | TNFR1 / CD120a / TNFRSF1A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16045321
|
enQuireBio™
QP8566-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16065321
|
enQuireBio™
QP8566-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16055321
|
enQuireBio™
QP8566-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16075321
|
enQuireBio™
QP8566-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16085321
|
enQuireBio™
QP8566-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16095321
|
enQuireBio™
QP8566-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001056.1 | |
TNFR1 / CD120a / TNFRSF1A Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
7132 | |
Research Use Only | |
TNFRSF1A | |
Recombinant Protein | |
E. coli | |
VPHLGDREKR DSVCPQGKYI HPQNNSICCT KCHKGTYLYN DCPGPGQDTD CRECESGSFT ASENHLRHCL SCSKCRKEMG QVEISSCTVD RDTVCGCRKN QYRHYWSENL FQCFNCSLCL NGTVHLSCQE KQNTVCTCHA GFFLRENECV SCSNCKKSLE CTKLCLPQIE NVKGTEDSGT | |
Greater than 90% as determined by SDS-PAGE. |