missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TNFRSF10C Protein
A cDNA sequence encoding the TNFRSF10C was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13767-5ug
Additional Details : Weight : 0.01000kg
Specifications
8794 | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSATTARQE EVPQQTVAPQ QQRHSFKGEE CPAGSHRSEH TGACNPCTEG VDYTNASNNE PSCFPCTVCK SDQKHKSSCT MTRDTVCQCK EGTFRNENSP EMCRKCSRCP SGEVQVSNCT SWDDIQCVEE FGANATVETP AAEETMNTSP GTPAPAAEET MNTSPGTPAP AAEETMTTSP GTPAPAAEET MTTSPGTPAP AAEETMTTSP GTPA | |
Greater than 90.0% as determined by SDS-PAGE. |
TNFRSF10C Protein | |
Research Use Only | |
TNFRSF10C | |
Human | |
His | |
TNFRSF10C protein solution (1 mg/ml) containing Phosphate buffered saline (pH 7.4) and 10% glycerol. |