missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TNFSF10 / TRAIL / APO-2L Protein
A DNA sequence encoding the Homo sapiens (Human) TNFSF10 / TRAIL / APO-2L, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001177871.1 |
---|---|
Gene ID (Entrez) | 8743 |
Name | TNFSF10 / TRAIL / APO-2L Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16036050
|
enQuireBio™
QP6817-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16056050
|
enQuireBio™
QP6817-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16046050
|
enQuireBio™
QP6817-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16066050
|
enQuireBio™
QP6817-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16076050
|
enQuireBio™
QP6817-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16086050
|
enQuireBio™
QP6817-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001177871.1 | |
TNFSF10 / TRAIL / APO-2L Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
8743 | |
Research Use Only | |
TNFSF10 | |
Recombinant Protein | |
E. coli | |
TNELKQMQDK YSKSGIACFL KEDDSYWDPN DEESMNSPCW QVKWQLRQLV RKMILRTSEE TISTVQEKQQ NISPLVRERG PQRVAAHITG TRGRSNTLSS PNSKNEKALG RKINSWESSR SGHSFLSNLH LRNGELVIHE KGFYYIYSQT YFRFQEEIKE NTKNDKQMVQ YIYKYTSYPD PILLMKSARN SCWSKDAEYG LYSIYQGGIF ELKENDRIFV SVTNEHLIDM DHEASFFGAF LVG | |
Greater than 90% as determined by SDS-PAGE. |