missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TPI1 Protein

A cDNA sequence encoding the TPI1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13793-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15984719

Quantity Price
1 114.77 € / 5µg
10 107.33 € / 5µg
Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

7167
Greater than 95% as determined by SDS-PAGE.
Research Use Only
TPI1
Human
His
The TPI1 protein solution (0.5 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0), 1mM DTT and 10% glycerol.
TPI1 Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MAPSRKFFVG GNWKMNGRKQ SLGELIGTLN AAKVPADTEV VCAPPTAYID FARQKLDPKI AVAAQNCYKV TNGAFTGEIS PGMIKDCGAT WVVLGHSERR HVFGESDELI GQKVAHALAE GLGVIACIGE KLDEREAGIT EKVVFEQTKV IADNVKDWSK VVLAYEPVWA IGTGKTATPQ QAQEVHEKLR GWLKSNVSDA VAQSTRIIYG GSVTGATCKE LASQPDVDGF LVGGASLKPE FVDIINAKQ
Videos
SDS
Documents
Certificates