missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Transferrin Protein

A cDNA sequence encoding the Transferrin was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10893-1gr

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15985488

Quantity Price
1 445.65 € / 1g
6 419.36 € / 1g
Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

7018
Purity as determined by SDS-PAGE is 95%.
Research Use Only
TF
Recombinant Protein
Native Protein
MRLAVGALLV CAVLGLCLAV PDKTVRWCAV SEHEATKCQS FRDHMKSVIP SDGPSVACVK KASYLDCIRA IAANEADAVT LDAGLVYDAY LAPNNLKPVV AEFYGSKEDP QTFYYAVAVV KKDSGFQMNQ LRGKKSCHTG LGRSAGWNIP IGLLYCDLPE PRKPLEKAVA NFFSGSCAPC ADGTDFPQLC QLCPGCGCST LNQYFGYSGA FKCLKDGAGD VAFVKHSTIF ENLANKADRD QYELLCLDNT RKPVDEYKDC HLAQVPSHTV VARSMGGKED LIWELLNQAQ EHFGKDKSKE FQLFSSPHGK DLLFKDSAHG FLKVPPRMDA KMYLGYEYVT AIRNLREGTC PEAPTDECKP VKWCALSHHE RLKCDEWSVN SVGKIECVSA ETTEDCIAKI MNGEADAMSL DGGFVYIAGK CGLVPVLAEN YNKSDNCEDT PEAGYFAIAV VKKSASDLTW DNLKGKKSCH TAVGRTAGWN IPMGLLYNKI NHCRFDEFFS EGCAPGSKKD SSLCKLCMGS GLNLCEPNNK EGYYGYTGAF RCLVEKGDVA FVKHQTVPQN TGGKNPDPWA KNLNEKDYEL LCLDGTRKPV EEYANCHLAR APNHAVVTRK DKEACVHKIL RQQQHLFGSN VTDCSGNFCL FRSETKDLLF RDDTVCLAKL HDRNTYEKYL GEEYVKAVGN LRKCSTSSLL EACTFRRP
Transferrin Protein
1 g
< 1.0 EU per ug protein as determined by the LAL method.
One mg of Recombinant Human Transferrin will bind to approximately 2 micrograms of Fe.
Human
Untagged
The protein (1 mg/ml) was lyophilized with no additives.
Videos
SDS
Documents
Certificates